missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, 10 to 18 mesh

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

Supplier:  Thermo Scientific Chemicals 264750050

Quantity 5 kg
Packaging Plastic drum

 View more versions of this product

Catalog No. AC264750050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
Specifications

Chemical Identifiers

70955-01-0
440.42
FJUZEZRZKVMAMD-UHFFFAOYNA-N
Molecular sieves
C20H25FN2O8
MFCD00131613
73906282
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Beige
5 kg
680 to 760g/L
> 20% (50% rH, 20°C, 24 h)
70955-01-0
MFCD00131613
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
440.42
Molecular sieves 4A
Plastic drum
< 1.5% (550°C, 2 hrs.)
Granules
C20H25FN2O8
FJUZEZRZKVMAMD-UHFFFAOYNA-N
Molecular sieves
73906282
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

SDS
Documents

Documents

Promotions

Promotions