Learn More
Molecular sieves, 4A, 0.4-0.8mm (0.02-0.03in) beads
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Supplier: Thermo Scientific Chemicals L055120B
| Quantity | 1000 g |
|---|
Description
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads are useful in general gas drying applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 70955-01-0 | |
| 440.42 | |
| FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
| (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
| C20H25FN2O8 | |
| MFCD00131613 | |
| 73906282 | |
| COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Specifications
| Molecular sieves, 4A | |
| Hygroscopic | |
| 70955-01-0 | |
| MFCD00131613 | |
| FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
| (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
| 73906282 |
| 1000 g | |
| 0.4 to 0.8mm (0.02 to 0.03 in) beads | |
| C20H25FN2O8 | |
| Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone. | |
| COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
| 440.42 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures