missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 4 Å, 8 to 12mesh, Honeywell Fluka™

Beads, 8-12 mesh

$742.61 - $1,762.47

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
6002078
View Documents Promotion Details
Honeywell-Fluka
2086041KG
1 kg
Each for $742.61
Only null left
 
6002080
View Documents Promotion Details
Honeywell-Fluka
2086045KG
5 kg
Each for $1,762.47
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

70955-01-0
440.42
FJUZEZRZKVMAMD-UHFFFAOYNA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
C20H25FN2O8
MFCD00131613
73906282
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

8 to 12mesh
C20H25FN2O8
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
73906282
70955-01-0
Na12[(AlO2)12(SiO2)12] · xH2O
NONH for all modes of transport
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
440.42
Molecular Sieves, 4 Å
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

H315-H319-H335

SDS
Documents

Documents