missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 13X, 8 to 12mesh, Honeywell Fluka™
Beads, 8-12 mesh
$2,174.43 - $2,174.43
Chemical Identifiers
| CAS | 63231-69-6 |
|---|---|
| MDL Number | MFCD00131613 |
| IUPAC Name | Molecular sieves 13X |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 63231-69-6 | |
| MFCD00131613 | |
| Molecular sieves 13X |
Specifications
| 8 to 12mesh | |
| 63231-69-6 | |
| MFCD00131613 | |
| Molecular sieves 13X |
| 5 kg | |
| Na86[AlO2)86(SiO2)106] · xH2O | |
| NONH for all modes of transport | |
| Molecular Sieves, 13X |
Safety and Handling
P261-P305 + P351 + P338
H315-H319-H335