missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 13X, 8 to 12mesh, Solstice

Catalog No. 6002082
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002082 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002082 Supplier Solstice Supplier No. 2086475KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beads, 8-12 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 5 kg
Mesh Size 8 to 12mesh
Linear Formula Na86[AlO2)86(SiO2)106] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular Sieves, 13X
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335