missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 1.6-2.5 mm (0.063-0.098in) beads

CAS: 63231-69-6

$88.99 - $799.68

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2110930
View Documents
Thermo Scientific Chemicals
B2110930
250 g
Each for $88.99
Only null left
 
AAB211090B
View Documents
Thermo Scientific Chemicals
B211090B
1000 g
Each for $231.26
Only null left
 
AAB211090I
View Documents
Thermo Scientific Chemicals
B211090I
5000 g
Each for $799.68
Only null left
 
Description

Description

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves 13X

Specifications

Hygroscopic
63231-69-6
Insoluble in water.
Molecular sieves, 13X, 1.6 to 2.5mm (0.063 to 0.098 in) beads
250 g
MFCD00131613
Molecular sieves 13X
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only