missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 1.6-2.5 mm (0.063-0.098in) beads

CAS: 63231-69-6

Supplier:  Thermo Scientific Chemicals B211090I

 View more versions of this product

Catalog No. AAB211090I


Only null left
Explore available promotions

Description

Description

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hygroscopic
Molecular sieves, 13X
63231-69-6
Insoluble in water.
5000 g
1.6 to 2.5mm (0.063 to 0.098 in) beads
MFCD00131613
Molecular sieves 13X

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves 13X
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only