missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ X-Gal (5-Bromo-4-Chloro-3-Indolyl β-D-Galactopyranoside)

Catalog No. B1690
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1690 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1690 Supplier Invitrogen™ Supplier No. B1690
  • ONLINE EXCLUSIVE PRICE
    $324.00
    $265.68 / Each

    Save $58.32

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Widely used chromogenic substrate for β-galactosidase

Substrate yields a dark blue precipitate at the site of enzymatic activity. X-Gal is useful for numerous histochemical and molecular biology applications, including detection of lacZ activity in cells and tissues. In contrast to β-glucuronidase as a gene marker, β-galactosidase can be fixed in cells and tissues with glutaraldehyde without loss of activity and detected with high resolution using X-Gal.

Additional Information About Glycosidase Enzymes
Glycosidase enzymes exhibit very high selectivity for hydrolysis of their preferred sugars. For example, β-galactosidase rapidly hydrolyzes β-D-galactopyranosides but usually does not hydrolyze either the anomeric α-D-galactopyranosides or the isomeric β-D-glucopyranosides. Endogenous glycosidase activity is frequently used to characterize strains of microorganisms and to selectively label organelles of mammalian cells; defects in glycosidase activity are characteristic of several diseases.

In addition, glycosidases are important reporter gene markers. Specifically, lacZ, which encodes β-galactosidase, is extensively used as a reporter gene in animals and yeast, whereas the β-glucuronidase (GUS) gene is a popular reporter gene in plants. Glycosidase substrates are also used in conjunction with glycosidase-conjugated secondary detection reagents in immunohistochemical techniques and enzyme-linked immunosorbent assays (ELISAs).

  • Chromogenic substrate
  • Cell permeant
  • β-galactosidase activity can still be detected using X-Gal after fixation

Beta-Galactosidase Assays, Cell Analysis, Cellular Imaging, Cloning, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunohistochemistry (IHC), Proteins, Expression, Isolation and Analysis, Reporter Assays, Transformation

Order Info

Shipping Condition: Room Temperature

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Colorimetric
For Use With (Application) Molecular Biology
Quantity 1 g
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Substrate Type Beta-Gal Substrate
Shipping Condition Room Temperature
How do I resuspend IPTG and X-Gal?

IPTG can be reconstituted in water. Make a stock of 100 mM in water and store working aliquots at -20°C. X-gal can be reconstituted in DMSO, or in a 50:50 mix of DMSO and water. To do the latter, you must dissolve in DMSO first, and then add water to bring up to final volume. It is not necessary to filter sterilize these solutions.

The X-gal solution should be protected from light. To make plates, add 50 ug/ml X-gal and 1 mM (0.24 mg/mL) IPTG to LB/agar that has been cooled down to 50°C. To spread on top of plates, use 50 µl 2% stock of X-gal and 30 µl 100 mM stock of IPTG. 

For Research Use Only. Not for use in diagnostic procedures.