missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vancomycin, BODIPY™ FL Conjugate (BODIPY™ FL Vancomycin)

Catalog No. V34850
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V34850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V34850 Supplier Invitrogen™ Supplier No. V34850

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BODIPY™ FL vancomycin, which contains a single BODIPY™ dye per vancoymycin molecule, is a green-fluorescent analog of this important antibiotic, which is active against gram-positive bacteria, including entereococci.

BODIPY™ FL vancomycin, which contains a single BODIPY™ dye per vancoymycin molecule, is a green-fluorescent analog of this important antibiotic, which is active against gram-positive bacteria, including entereococci. Although its biological activity has not been reported, it may be useful for detecting vancomycin binding sites and transport, for fluorescence polarization assays, and for the study and detection of vancomycin-resistant enterococci.

To make a concentrated stock solution, dissolve in either high quality anhydrous DMSO (1 mM) or H2O (∼60 mM). Store DMSO stock solutions frozen, desiccated and protected from light. Aqueous stock solutions should be used fresh as prolonged storage may result in aggregation.

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 100 μg
Shipping Condition Room Temperature
Product Line BODIPY
Label Type BODIPY Dyes
Molecular Formula C80H88BF2Cl2N11O25
Physical Form Solid
Product Type Antibiotic
Excitation/Emission 504/511 nm
Molecular Weight (g/mol) 1723.35

For Research Use Only. Not for use in diagnostic procedures.