missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMA-DPH (1-(4-Trimethylammoniumphenyl)-6-Phenyl-1,3,5-Hexatriene p-Toluenesulfonate)

Catalog No. T204
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T204 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T204 Supplier Invitrogen™ Supplier No. T204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with fluorescence microscopy

TMA-DPH contains a cationic trimethylammonium substitute that acts as a surface anchor to improve the localization of the fluorescent probe of membrane interiors, DPH.

Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, Membranes (General) & Lipids

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 25 mg
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Plasma Membrane
Detection Method Fluorescence
Molecular Formula C28H31NO3S
Physical Form Solid
Color Violet
Product Type Cell Surface Anchor

For Research Use Only. Not for use in diagnostic procedures.