missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thapsigargin

Catalog No. p-4921138
Encompass
Change view
Click to view available options
Quantity:
1 mg
20 x 50 μg
Catalog No. Quantity
T7458 1 mg
T7459 20 x 50 μg
2 options

Catalog No. T7458

Supplier: Invitrogen™ T7458

Only null left

Tumor promoter

Thapsigargin is a naturally occurring sesquiterpene lactone isolated from the umbelliferous plant Thapsia garganica. This tumor promoter releases Ca2++ from intracellular stores by specifically inhibiting the endoplasmic reticulum Ca2++-ATPase. It does not directly affect plasma membrane Ca2++-ATPases, Ins 1,4,5-P3 production or protein kinase C activity.

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Label Type Unlabeled
Molecular Formula C34H50O12
Physical Form Solid
Product Type Inhibitor

For Research Use Only. Not for use in diagnostic procedures.