missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TC-FlAsH™ TC-ReAsH™ II In-Cell Tetracysteine Tag Detection Kit, with Mammalian TC-Tag Gateway™ Expression Vectors (Green Fluorescence) (Red Fluorescence)

Catalog No. T34563
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34563 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34563 Supplier Invitrogen™ Supplier No. T34563

Includes

The kit contains FlAsH-EDT2 labeling reagent, ReAsH-EDT2 labeling reagent, BAL wash buffer, pcDNA6.2/cTC-Tag-DEST, pcDNA6.2/nTC-Tag-DEST, and pcDNA6.2/nTC-Tag-p64 Control Plasmid.

An expression tag-based fluorescence labeling system for live-cell labeling

The expression construct is created when the Gateway™ entry clone (containing the gene of interest) is recombined with one of two destination vectors (pcDNA6.2/nTC-Tag-DEST for N-terminal tagging or pcDNA6.2/cTC-Tag-DEST for C-terminal tagging). The expression construct is then used to transform the host strain. The protein can be detected either with green-fluorescent FlAsH-EDT2 reagent or red-fluorescent ReAsH-EDT2 reagent. The tag is very small (6 amino acids) and the protein of interest is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Labeling Chemistry, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

Order Info

Shipping Condition: Wet Ice

Specifications

Product Type Tetracysteine Tag Detection Kit
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Assay Tetracysteine Tag Detection
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Color Red, Green
Label or Dye FlAsH, ReAsH
Shipping Condition Wet Ice
Format Kit
Product Line Gateway, TC-FlAsH II, TC-ReAsH II
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.