missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Resorufin β-D-Galactopyranoside

Catalog No. r1159
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1159 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1159 Supplier Invitrogen™ Supplier No. R1159

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the hydrolysis product, resorufin

The fluorogenic β-galactosidase substrate resorufin β-D-galactopyranoside yields the hydrolysis product, resorufin, with absorption/emission maxima at 571/585nm.

Beta-Galactosidase Assays, Cell Analysis, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Quantity 25 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 571/585
Shipping Condition Room Temperature
What is the excitation/emission maxima of resorufin?

The excitation/emission maxima is 563/587 nm.

For Research Use Only. Not for use in diagnostic procedures.