missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PFB-FDGlu (5-(Pentafluorobenzoylamino)Fluorescein Di-β-D-Glucopyranoside)

Catalog No. p11947
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11947 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11947 Supplier Invitrogen™ Supplier No. P11947

PFB-FDGlu is a cell-permeable substrate used for live cell imaging of GCase activity. It fluoresces green upon cleavage by lysosomal GCase, facilitating specific, single-cell analysis via high-content imaging, flow cytometry, and microscopy.

PFB-FDGlu (5-(Pentafluorobenzoylamino) Fluorescein Di-beta-D-Glucopyranoside) is a glycosidase activity probe for live cell imaging of lysosomal glucocerebrosidase (GCase) activity. This enzyme is vital for glycolipid metabolism, and its deficiency is linked to diseases like Gaucher’s and Parkinson's. PFB-FDGlu fluoresces green upon cleavage by GCase, allowing specific, single-cell analysis via flow cytometry, fluorescent microscopy, and high-content imaging.

PFB-FDGlu is cell-permeable and when loaded into cells, localizing to endosomal and lysosomal compartments while maintaining cell viability. Upon enzymatic cleavage by active GCase enzymes, it yields a green-fluorescent dye (PFB-F) with excitation/emission maxima of approximately 492/516 nm. This fluorescence can be detected using fluorescence microscopy, high-content imaging, and flow cytometry, allowing for specific, single-cell analysis of GCase activity.

Advantages of PFB-FDGlu for detecting GCase activity in lysosomes:

  • High specificity—utilizes a lysosomal GCase substrate for measurement of GCase activity, ensuring specific lysosomal staining (note that with long incubations the fluorescent PFB-F will migrate to the cytoplasm)
  • Live cell imaging—allows for the assessment of GCase activity in intact, living cells with minimal impact on cell viability, even with extended incubation
  • Single-cell analysis—enables specific, single-cell analysis with flow cytometry, high-content imaging, and microscopy to distinguish between different cell types
  • Bright fluorescence—yields a bright green fluorescent dye upon cleavage by GCase, enhancing detection sensitivity
  • Versatile protocol—can be combined with other phenotypic indicators, such as viability stains and antibodies, to identify distinct cell populations or as a selection criterion for cell sorting

GCase, encoded by the GBA1 gene, is a lysosomal hydrolase that converts glucosylceramide to glucose and ceramide. Mutations in GBA1 leading to GCase deficiency can cause lysosomal storage disorders. GCase deficiencies are implicated in several diseases, including Gaucher's disease and Parkinson's disease.

Order Info

Shipping Condition: Room Temperature

Specifications

Cell Permeability Cell-Permeant
Color Green
Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 5 mg
Substrate GCase Substrate
Substrate Properties Chemical Substrate
Substrate Type Beta-Glucosidase Substrate
Target Enzyme Beta-Glucosidase
Excitation/Emission 492/526
For Use With (Equipment) Flow Cytometer, High Content Imaging Instrument, Fluorescent Microscope, Confocal Microscope
Label or Dye Fluorescein
Product Type Substrate
Shipping Condition Room Temperature
Storage Requirements Freezer (-5°C to -30°C)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.