missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PED-A1 (N-((6-(2,4-DNP)Amino)Hexanoyl)-1-(BODIPY™ FL C5)-2-Hexyl-Sn-Glycero-3-Phosphoethanolamine) (Phospholipase A1 Selective Substrate)

Catalog No. A10070
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10070 Supplier Invitrogen™ Supplier No. A10070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PED-A1-labeled phospholipid

Molecular Probes phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells.

  • Label (Ex/Em): PED-A1
  • Specificity: PLA1
  • Cleavage product: BODIPY™ FL C5
  • Lyophilized product can be dissolved in DMSO for use
  • Fluorescence is measured either in microplate assays or using flow cytometry
  • Cell-Permeant
Note: Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Solid
Product Line BODIPY
Quantity 100 μg
Substrate Phospholipase A1 selective substrate
Substrate Properties Lipid-Based Substrate
Target Enzyme Phospholipase
For Use With (Equipment) Flow Cytometer, Microplate Reader
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.