missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nigericin, Free Acid
Description
For intracellular calibration of fluorescence response of cytosolic pH indicators
Intracellular calibration of the fluorescence response of cytosolic pH indicators is typically performed using the K+/H+ ionophore nigericin, which causes equilibration of intracellular and extracellular pH in the presence of a depolarizing concentration of extracellular K+.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 10 mg |
| Shipping Condition | Room Temperature |
| Indicator | Ionophore |
| Molecular Formula | C40H68O11 |
| Physical Form | Solid |
| Product Type | Potassium Ionophore |
For Research Use Only. Not for use in diagnostic procedures.