missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBT/BCIP Reagent Kit

Catalog No. N6547
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N6547 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N6547 Supplier Invitrogen™ Supplier No. N6547

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

1 g each of NBT and BCIP

For chromogenic detection of phosphatase.

The NBT/BCIP Reagent Kit includes 1 g samples of both NBT and BCIP for the chromogenic detection of phosphatase.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Absorbance
Quantity 1 Kit
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.