missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUP (4-Methylumbelliferyl Phosphate, Free Acid)

Catalog No. m6491
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M6491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M6491 Supplier Invitrogen™ Supplier No. M6491

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MUP is a fluorescent substrate for alkaline phosphatases

Alkaline phosphatase mediated hydrolysis of its phosphate substituent yields the blue-fluorescent 4-methylumbelliferyl (excitation/emission approx. 386/448nm).

Alkaline Phosphatase Assays, Biochemical Toxicology Assays, Cell Analysis, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target and Lead Identification and Validation, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 1 g
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Alkaline Phosphatase
Excitation/Emission 360⁄449
For Use With (Equipment) Fluorometer, Microplate Reader
Label or Dye MUP (4-methylumbelliferyl phosphate)
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.