missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Methotrexate, Fluorescein, triammonium Salt (Fluorescein Methotrexate)

Catalog No. M1198MP
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1198MP 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1198MP Supplier Invitrogen™ Supplier No. M1198MP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To probe DHFR expression

The green-fluorescent fluorescein methotrexate (absorption/emission maxima approx. 496/516nm) can be used to probe dihydrofolate reductase (DHFR) expression by fluorescence microscopy or flow cytometry.

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Label Type Classic Dyes
Molecular Formula C46H54N14O9S
Physical Form Solid
Product Type Fluorescein Methotrexate

For Research Use Only. Not for use in diagnostic procedures.