missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mag-Fura-2, AM, cell permeant

Intracellular magnesium indicator that is ratiometric and UV light-excitable

$932.00

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Magnesium Indicator
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M1292
Encompass
View Documents Promotion Details
Invitrogen™
M1292
20 x 50 μg
Each for $932.00
Only null left
 
Description

Description

  • Acetoxymethyl (AM) ester form
  • Useful for noninvasive intracellular loading

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Magnesium Detection

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Cell Viability and Proliferation
Magnesium Indicator
Room Temperature
Fluorescence
Fluorescence Microscope, Microphotometer, Flow Cytometer
Fluorescent Dye-Based
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.