missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ionomycin, Calcium Salt

Catalog No. I24222
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I24222 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I24222 Supplier Invitrogen™ Supplier No. I24222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ionomycin is an effective Ca2+ ionopohre that is commonly used to both modify intracellular Ca2+ concentrations and to calibrate fluorescent Ca2+ indicators when studying the regulatory properties of Ca2+ in cellular processes.

Ionomycin is an effective Ca2+ ionopohre that is commonly used to both modify intracellular Ca2+ concentrations and to calibrate fluorescent Ca2+ indicators when studying the regulatory properties of Ca2+ in cellular processes. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations, at efficiencies that are greater than or equal to those for Ca2+.

To make a concentrated stock solution, dissolve in either high quality anhydrous DMSO (1.6 mg/mL) or 100% ethanol (20 mg/mL). Store frozen, desiccated and protected from light.

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Indicator Ionophore
Molecular Formula C41H70CaO9
Physical Form Solid
Product Type Calcium Ionophore
Molecular Weight (g/mol) 747.08

For Research Use Only. Not for use in diagnostic procedures.