missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FR-31564 (Fosmidomycin, Sodium Salt)

Catalog No. F23103
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F23103 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F23103 Supplier Invitrogen™ Supplier No. F23103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blocks a pathway of isoprene synthesis

The phosphonate antibiotic FR-31564 (fosmidomycin) is an effective antimalarial agent that functions by blocking a mevalonate-independent pathway of isoprene synthesis. The antibiotic activity of FR-31564 is potentiated by glucose 1-phosphate. FR-31564 is also active against a variety of gram-negative bacteria.

Analysis of Bacteria, Analysis of Viruses, Fungi and Parasites, Antibiotic Binding Studies, Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Microbiological Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 25 mg
Shipping Condition Room Temperature
Molecular Formula C4H9NNaO5P
Physical Form Powder
Product Type Antibiotic
How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

For Research Use Only. Not for use in diagnostic procedures.