missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FM™ 4-64 Dye (N-(3-Triethylammoniumpropyl)-4-(6-(4-(Diethylamino) Phenyl) Hexatrienyl) Pyridinium Dibromide)

Lipophilic dye

$734.00 - $860.00

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Detection Method Fluorescence
Product Line FM
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
T13320
Encompass
View Documents Promotion Details
Invitrogen™
T13320
10 x 100μg
Each for $860.00
Only null left
 
T3166
Encompass
View Documents Promotion Details
Invitrogen™
T3166
1mg
Each for $734.00
Only null left
 
Description

Description

The styryl dye FM 4-64 has been reported to selectively stain yeast vacuolar membranes with red fluorescence (excitation/emission maxima ∽515/640nm). This dye is an important tool for visualizing vacuolar organelle morphology and dynamics, for studying the endocytic pathway and for screening and characterizing yeast endocytosis mutants. FM 4-64 is also available as 1mg in a single vial (T-3166).

Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, General Cell Tracing, Microbial Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Red
Fluorescence Microscope
FM
Fluorescent Dye
Plasma Membrane
Store at room temperature and protect from light.
Fluorescence
Room Temperature
Dye
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.