missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein Di-β-D-Glucuronide (FDGlcU)
Description
Fluorescein di-β-D-glucuronide (FDGlcU) is colorless and nonfluorescent until it is hydrlozed to the monoglucuronide and then to the highly fluorescent fluorescein.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Permeability | Cell-permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Quantity | 5 mg |
| Substrate | Beta-Glucuronidase (GUS) Substrate |
| Substrate Properties | Chemical Substrate |
| Excitation/Emission | 490/514 |
| Shipping Condition | Room Temperature |
| Excitation Wavelength Range | 490 nm |
| Emission Wavelength Range | 517 nm |
Frequently Asked Questions (FAQs)
Can I make a higher stock solution of Fluorescein Di-β-D-Glucuronide (FDGlcU) (Cat. No. F2915) e.g., 50 mM in PBS?
Unfortunately, we do not have data on the upper limit of solubility for this reagent in PBS. This is something you would need to test empirically.br/>
What is the molecular weight of Fluorescein Di-β-D-Glucuronide (FDGlcU) (Cat. No. F2915)?
The molecular weight of the above product is 684.56 g/mol.br/>
For Research Use Only. Not for use in diagnostic procedures.