missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluorescein Di-β-D-Glucopyranoside (FDGlu)
Description
Fluorescein di-β-D-glucopyranoside is one of the most sensitive substrates for glucosidases. The nonfluorescent substrate is sequentially hydrolyzed by β-glucosidase, first to fluorescein monoglucoside and then to highly fluorescent fluorescein. Enzyme-mediated hydrolysis of fluorescein di-β-D-glucopyranoside can be followed by the increase in either absorbance or fluorescence.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Permeability | Cell-permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| For Use With (Application) | Glucosidases Substrate |
| Quantity | 5 mg |
| Substrate | Beta-Glucosidase Substrate |
| Substrate Properties | Chemical Substrate |
| Excitation/Emission | 490/514 |
| Shipping Condition | Room Temperature |
| Excitation Wavelength Range | 490 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.