missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Di-β-D-Galactopyranoside (FDG)

Catalog No. F1179
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1179 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1179 Supplier Invitrogen™ Supplier No. F1179

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell permeant

Fluorescein di-β-D-galactopyranoside (FDG) is one of the most sensitive substrates for galactosidases. Nonfluorescent FDG is sequentially hydrolyzed by β-galactosidase, first to fluorescein monogalactoside (FMG) and then to highly fluorescent fluorescein. Enzyme-mediated hydorlysis of FDG can be followed by the increase in either absorbance or fluorescence.

Order Info

Shipping Condition: Room Temperature

Specifications

Cell Permeability Cell-permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
For Use With (Application) Galactosidase
Quantity 5 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 490/514
Shipping Condition Room Temperature
Excitation Wavelength Range 490 nm
Emission Wavelength Range 517 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.