missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein-5-(and-6)-Sulfonic Acid, Trisodium Salt

Catalog No. F1130
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1130 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1130 Supplier Invitrogen™ Supplier No. F1130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell impermeant

The fluorescein-5-(and-6)-sulfonic acid can be used as a non-fixable polar tracer to investigate fusion, lysis and gap-junctional communication.

Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, General Cell Tracing, Ionic Homeostasis & Signaling, Neuronal Tracing, pH Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 100 mg
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Cell Membranes & Lipids
Detection Method Fluorescence
Molecular Formula C20H9Na3O8S
Physical Form Solid
Color Green
Product Type Cell Tracer

For Research Use Only. Not for use in diagnostic procedures.