missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoReporter™ lacZ/Galactosidase Quantitation Kit DFS Item

Reaction produces blue fluorescent product

Supplier:  Invitrogen™ F2905

Catalog No. F2905


Only null left
Explore available promotions

Description

Description

The FluoReporter™ lacZ/Galactosidase Quantitation Kit includes the β-galactosidase substrate, 3-carboxyumbelliferyl β-D-galactopyranoside that produces the blue-fluorescent hydrolysis product, 7-hydroxycoumarin-3-carboxlic acid.

Beta-Galactosidase Assays, Cell Analysis, Enzyme and Protein Activity Assays, Flow Cytometry, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Beta-Galactosidase Reporter Gene Assay System
β-Gal (lacZ) Assay
Room Temperature
Beta-Gal Substrate
Beta-Galactosidase
FluoReporter
Fluorescence
Store in freezer (-5 to -30°C) and protect from light.
CUG
Chemical Substrate
Kit
1 Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.