missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DQ™ Gelatin From Pig Skin, Fluorescein Conjugate - Special Packaging

Catalog No. d12054
Encompass
Change view
Click to view available options
Quantity:
5 x 1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12054 5 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12054 Supplier Invitrogen™ Supplier No. D12054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analog of natural substrate

Our DQ gelatin is a fluorogenic substrate that can be used to detect protease activity in vitro with high sensitivity. This substrate consists of highly quenched, fluorescein-labeled gelatin. Upon proteolytic digestion, its bright, green fluorescence is revealed and can be used to measure enzymatic activity. The fluorescence increase can be monitored with a fluorescence microplate reader or fluorometer.

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Form Powder
Product Line DQ
Quantity 5 x 1 mg
Substrate Protease Substrate
Substrate Properties Protein-Based Substrate
Target Enzyme Metalloproteinase
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.