missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPX (p-Xylene-Bis-Pyridinium Bromide)

Catalog No. X1525
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X1525 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X1525 Supplier Invitrogen™ Supplier No. X1525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cationic quencher

DPX (p-xylene-bis-pyridinium bromide) can be used in membrane fusion or permeability assays with the polyanionic, polar tracer dye ANTS (A350).

Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 g
Shipping Condition Room Temperature
Molecular Formula C18H18Br2N2
Physical Form Solid
Product Type DPX Quencher

For Research Use Only. Not for use in diagnostic procedures.