missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Catalog No. p-4921019
Encompass
Change view
Click to view available options
Quantity:
5mg
10 x 10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6567 5mg
D22065 10 x 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D6567 Supplier Invitrogen™ Supplier No. D6567

Continuously assay acid phosphatases at low pH

DiFMUP is a fluorinated 4-methylumbelliferyl phosphate (MUP) derivative that has a lower pKa than that of MUP, making DiFMUP an excellent substrate for continuously assaying acid phosphatases at low pH. The reaction product of DiFMUP has excitation/emission maxima of approx. 358/450nm.

Biochemical Kinase Assays, Biochemical Toxicology Assays, Cell Analysis, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Kinase Biology, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

Specifications

Color Blue
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 5mg
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Phosphatases
Excitation/Emission 358⁄455
For Use With (Equipment) Microplate Reader
Label or Dye DiFMU
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.