missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dexamethasone Fluorescein

Catalog No. D1383
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1383 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1383 Supplier Invitrogen™ Supplier No. D1383
  • ONLINE EXCLUSIVE PRICE
    $266.00
    $218.12 / Each

    Save $47.88

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

missing translation for 'validMainframeMsg'

The synthetic steroid hormone dexamethasone binds to the glucocorticoid receptor, producing a steroid-receptor complex that then localizes in the nucleus and regulates gene transcription.

The synthetic steroid hormone dexamethasone binds to the glucocorticoid receptor, producing a steroid-receptor complex that then localizes in the nucleus and regulates gene transcription. The green fluorescent analog, fluorescein dexamethasone should be useful for studying the mechanism of glucocorticoid receptor activation, although at a slower rate than unmodified dexamethasone.

For stock concentrations up to 1 mM, DMF or aqueous buffers above pH 6 are recommended as diluent. Stock solutions can be stored as single-use aliquots, protected from light at -20°C.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 5 mg
Shipping Condition Room Temperature
Label Type Classic Dyes
Molecular Formula C45H45FN2O9S2
Physical Form Solid
Product Type Dexamethasone Fluorescein
Excitation/Emission 494/519 nm
Molecular Weight (g/mol) 840.98

For Research Use Only. Not for use in diagnostic procedures.