missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMAC, t-BOC-Leu-Met (7-Amino-4-Chloromethylcoumarin, t-BOC-L-Leucyl-L-Methionine amide)

Catalog No. A6520
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A6520 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6520 Supplier Invitrogen™ Supplier No. A6520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleavage produces blue-fluorescence

The CMAC peptidase substrate, t-BOC-Leu-Met can be used to measure peptidase activity in solution or in live cells. The substrate has an excitation/emission maxima approx. 330/403. After cleavage by peptidases, the product produces blue-fluorescence with excitation/emission maxima approx.351/430nm.

Apoptosis, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Quantity 5 mg
Substrate CMAC peptidase substrate
Substrate Properties Peptide-Based Substrate
Substrate Type Peptidase Substrate
Target Enzyme Calpain
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.