missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12FDG (5-Dodecanoylaminofluorescein Di-β-D-Galactopyranoside)

Catalog No. D2893
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2893 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2893 Supplier Invitrogen™ Supplier No. D2893
  • ONLINE EXCLUSIVE PRICE
    $950.00
    $779.00 / Each

    Save $171.00

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Produces a green fluorescence

The β-galactosidase substrate, C12FDG has been covalently modified to include a 12-carbon lipophilic moiety. Once inside the cell, the substrate is cleaved by β-galactosidase producing a fluorescent product that is well retained by the cells, probably by incorporation of the liphophilic tail within the cell membrane. C12FDG is the main component in the ImaGene™ Green kit.

Beta-Galactosidase Assays, Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Flow Cytometry, Flow Cytometry Antibodies and Secondary Detection, Glycosidase, Neuraminidase, and Related Enzyme Activity, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Quantity 5 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 490⁄514
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.