missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL Prazosin

Catalog No. b7433
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B7433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B7433 Supplier Invitrogen™ Supplier No. B7433

Localize and detect the a1-adrenergic receptor

Prazosin is a high-affinity antagonist for the a1-adrenergic receptor. The green-fluorescent BODIPY FL prazosin derivative can be used to localize and detect the a1-adrenergic receptor.

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Glutathione Detection, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 100 μg
Shipping Condition Room Temperature
Product Line BODIPY
Label Type BODIPY Dyes
Molecular Formula C28H32BF2N7O3
Product Type Antagonist

For Research Use Only. Not for use in diagnostic procedures.