missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BOCILLIN™ FL Penicillin, Sodium Salt

Catalog No. B13233
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13233 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13233 Supplier Invitrogen™ Supplier No. B13233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Penicillin-binding proteins, present on the cytoplasmic membranes of eubacteria, can be detected with BOCILLIN FL penicillin and BOCILLIN 650/665 penicillin.

Penicillin-binding proteins, present on the cytoplasmic membranes of eubacteria, can be detected with BOCILLIN FL penicillin and BOCILLIN 650/665 penicillin.

Dissolve in high-quality anhydrous DMSO or water for a stock concentration of 1 to 10 mM.

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Product Line BOCILLIN
Label Type Other Label(s) or Dye(s)
Molecular Formula C30H31BF2N5NaO6S
Physical Form Solid
Product Type Antibiotic
Excitation/Emission 504/511 nm
Molecular Weight (g/mol) 661.46

For Research Use Only. Not for use in diagnostic procedures.