missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bis-ANS (4,4'-Dianilino-1,1'-Binaphthyl-5,5'-Disulfonic Acid, Dipotassium Salt)

Catalog No. b153
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B153 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B153 Supplier Invitrogen™ Supplier No. B153

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Binds with affinity that is often orders-of-magnitude higher than 1-8-ANS as probe for nonpolar cavities in proteins

Anilinonaphthalene sulfonates (ANS) are essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes. This property makes it a sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.

Cell Analysis, Cell Structure, Cellular Imaging, Cytoskeleton, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids
Product Line Molecular Probes
Label Type Other Label(s) or Dye(s)
Molecular Formula C32H22K2N2O6S2
Physical Form Solid
Product Type Protein Folding Indicator

For Research Use Only. Not for use in diagnostic procedures.