missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Microplate Lysis Reagent

Catalog No. M10510
Encompass
Change view
Click to view available options
Volume:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume
M10510 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10510 Supplier Invitrogen™ Supplier No. M10510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One 50mL bottle

A buffered nonionic detergent solution for the lysis of adherent or nonadherent cultured cells under non-denaturing conditions

Simply remove the cell culture medium, add the Microplate Lysis Reagent, and incubate. The resulting cell lysates are suitable for many assays types, including the Amplex and EnzChek microplate assays.

  • Ready to use—after media removal the cells are lysed directly in the microplate
  • Validated for use with Amplex and EnzChek cell-based microplate assays
  • Gentle, nondenaturing lysis conditions

Microplate Lysis Reagent is a ready-to-use buffered nonionic detergent solution that is simply applied to adherent or nonadherent cells. After a five-minute incubation (optimization of this time may be required), a cell lysate is obtained that can be directly used in downstream microplate-based assays. Microplate Lysis Reagent is a superior choice for the Amplex and EnzChek micropate assays. Because of the gentle, nondenaturing conditions, Microplate Lysis Reagent can be adapted to many different cell types and downstream applications/assays.

Cell Lysis, Protein Sample Preparation & Protein Purification, Proteins, Expression, Isolation & Analysis

Specifications

Content And Storage Store at <25°C.
GC-Rich PCR Performance Medium
Product Line Amplex, EnzChek
Product Type Microplate Lysis Reagent
For Use With (Application) Real Time PCR (qPCR)
Reagent Type Cell Lysis Buffer
Volume 50 mL

For Research Use Only. Not for use in diagnostic procedures.