missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647 Decapack Set

Catalog No. A32757
Encompass
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32757 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A32757 Supplier Invitrogen™ Supplier No. A32757

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

In convenient single-use packaging

The Alexa Fluor™ 647 Reactive Dye Decapack is optimized for use with aminoallyl labeling techniques. The single-use packaging of the reactive dye eliminates the need to aliquot bulk dyes, providing convenience and ensuring maximum activity of the dyes. The specially packaged Alexa Fluor 647 dye is compatible with both aminoallyl dUTP- and aminoallyl UTP-modified nucleotides as well as most commercially available aminoallyl-based labeling kits. Use with aminoallyl labeling techniques.

Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Room Temperature

Specifications

Product Type Reactive Dye
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Alexa Fluor Dye
Labeling Method Indirect Labeling
Shipping Condition Room Temperature
Product Line Alexa Fluor
Quantity 1 Set

For Research Use Only. Not for use in diagnostic procedures.