missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Molecular Biology Reagents

Catalog No. p-319648
Change view
Click to view available options
Chemical Name or Material:
Saline Sodium Citrate Buffer
TAE Buffer
Tris Borate EDTA Buffer
Tris Hydrochloride Buffer
pH:
7 ±0.1
7.5 ±0.1
8.3 ±0.1
8.4 ±0.1
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material pH
MT46010CM TAE Buffer 8.3 ±0.1
MT46030CM Tris Hydrochloride Buffer 7.5 ±0.1
MT46011CM Tris Borate EDTA Buffer 8.4 ±0.1
MT46020CM Saline Sodium Citrate Buffer 7 ±0.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MT46010CM Supplier Corning™ Supplier No. 46010CM
Only null left

Used in molecular biology, nucleic acid purification applications

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

Specifications

Chemical Name or Material TAE Buffer
Concentration 10X
pH 8.3 ±0.1
Quantity 6 x 1 L
Sterility Sterile