missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Molecular Biology Reagents
Description
- Certificate of Analysis available
- Product can be diluted to obtain the desired concentration
Order Info
Shipped at ambient temperature
Specifications
Specifications
| Chemical Name or Material | Tris Borate EDTA Buffer |
| CAS | 10043-35-3 |
| Concentration | 10X |
| For Use With (Application) | Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis |
| pH | 8.4 ±0.1 |
| Physical Form | Liquid |
| Quantity | 6 x 1 L |
| Solution Type | Buffer |
| Sterility | Sterile |
| Grade | Molecular Biology |