Learn More
Moldex™ Rockets™ Reusable Ear Plugs
Rockets™ reusable ear plugs are designed to give a secure grip when handling so they easily slip into the ear. Triple-flange design provides maximum comfort for all-day wear.
$113.77 - $436.51
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
High Visibility No
Pairs per Package 50 Pairs/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Corded Plug Color Cord Material Packaging Type Price Quantity  
Catalog Number Mfr. No. Corded Plug Color Cord Material Packaging Type Price Quantity  
19-037-498
View Documents
Moldex™
MDX6405
Corded Purple Vinyl Pocket-Pak
Pack of 50 Pair for $205.22
Case of 4 Pack for $774.64
Only null left
19-160-0156
View Documents
Moldex™
6480
Uncorded Camouflage N/A Pocket-Pak
Case of 4 Each for $436.51
Only null left
19-037-497
View Documents
Moldex™
MDX6400
Uncorded Purple N/A Pocket-Pak
Pack of 50 Pair for $113.77
Case of 4 Pack for $429.44
Only null left
Description
Description
  • Easy grip handle
  • Air bubble in tip provides cushioned comfort
  • Can be washed and reused
  • 100% PVC-Free™
Specifications
Specifications
Yes
Universal
50 Pairs/Pk.
27 dB
Reusable
Class AL
No
TPE
Flanged
No
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'