Learn More
Moldex™ Rockets™ Reusable Ear Plugs
Catalog No. 191600156
Change view
Click to view available options
Corded:
Corded
Uncorded
Plug Color:
Camouflage
Purple
Cord Material:
N/A
Vinyl
Packaging Type:
Pocket-Pak
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Plug Color Cord Material Packaging Type
191600156 Uncorded Camouflage N/A Pocket-Pak
19037498 Corded Purple Vinyl Pocket-Pak
19037497 Uncorded Purple N/A Pocket-Pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191600156 Supplier Moldex™ Supplier No. 6480
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Rockets™ reusable ear plugs are designed to give a secure grip when handling so they easily slip into the ear. Triple-flange design provides maximum comfort for all-day wear.
  • Easy grip handle
  • Air bubble in tip provides cushioned comfort
  • Can be washed and reused
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
Cord Material N/A
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Military, Armed Forces, Federal and Local Law Enforcement, and Personal Use
High Visibility No
Packaging Type Pocket-Pak
Pairs per Package 50 Pairs/Pk.
Plug Material TPE
Plug Color Camouflage
Noise Reduction Rating (NRR) 27 dB
Corded Uncorded
Plug Shape Flanged
Product Type Reusable Earplugs
Disposable Reusable
Metal Detectable No
Dielectric Yes
Includes Pocket-size case
Show More Show Less