Learn More
Moldex™ Rockets™ Reusable Ear Plugs
Catalog No. 19037498
Change view
Click to view available options
Corded:
Corded
Uncorded
Plug Color:
Camouflage
Purple
Cord Material:
N/A
Vinyl
Packaging Type:
Pocket-Pak
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Plug Color Cord Material Packaging Type
19037498 Corded Purple Vinyl Pocket-Pak
191600156 Uncorded Camouflage N/A Pocket-Pak
19037497 Uncorded Purple N/A Pocket-Pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19037498 Supplier Moldex™ Supplier No. MDX6405
Only null left
Rockets™ reusable ear plugs are designed to give a secure grip when handling so they easily slip into the ear. Triple-flange design provides maximum comfort for all-day wear.
  • Easy grip handle
  • Air bubble in tip provides cushioned comfort
  • Can be washed and reused
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
Cord Color Purple
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
Packaging Type Pocket-Pak
Pairs per Package 50 Pairs/Pk.
Plug Material TPE
Plug Color Purple
Noise Reduction Rating (NRR) 27 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Includes Pocket-size case
Show More Show Less