Learn More
Thermo Scientific™ Modifying Enzymes
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Supplier: Thermo Scientific™ EL002AF
Description
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Features
- Convenient stand-alone enzymes and buffers for workflow optimization
- No animal-origin components in final product composition
- Reliable and reproducible results with rigorous quality testing
- Scalable and available as OEM products
Applications
- CRISPR/Cas-based genome editing
- Gene expression
- Generation of pluripotent cells
Specifications
| 30 U/μL | |
| 30 U/μL T4 RNA Ligase, 50 μL Store at -20°C. |
|
| Yes | |
| T4 RNA Ligase | |
| 70°C |
| RNA Ligation | |
| T4 RNA Ligase Reaction Buffer | |
| 1500 U | |
| RNA Research | |
| Liquid |
For Research Use Only. Not for use in diagnostic procedures.