missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modifying Enzymes

Catalog No. FEREO166AF
Change view
Click to view available options
Product Type:
DNase I
T4 RNA Ligase
T4 RNA Ligase 2
Vaccinia Capping Enzyme
Vaccinia mRNA Cap 2'-O-Methyltransferase
Quantity:
1000 U
1500 U
20,000 U
4000 U
5000 U
Concentration:
10 U/μL
30 U/μL
50 U/μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity Concentration
FEREO166AF Vaccinia Capping Enzyme 4000 U 10 U/μL
FEREM167AF Vaccinia mRNA Cap 2'-O-Methyltransferase 20,000 U 50 U/μL
FEREL002AF T4 RNA Ligase 1500 U 30 U/μL
FER4387661A T4 RNA Ligase 2 1000 U 10 U/μL
FEREN230AF DNase I 5000 U 50 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FEREO166AF Supplier Thermo Scientific™ Supplier No. EO166AF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells

Specifications

Concentration 10 U/μL
For Use With (Application) mRNA Capping
Content And Storage 10 U/μL Vaccinia Capping Enzyme, 400 μL

Store at -20°C.
Compatible Buffer 10X Capping Buffer
Quantity 4000 U
Product Type Vaccinia Capping Enzyme
Research Category RNA Research
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.