missing translation for 'onlineSavingsMsg'
Learn More

Plas Labs™ Model 855 Anaerobic Chambers

Use the versatile Plas Lab Model 855 Anaerobic Chamber as a controlled atmosphere or anaerobic chamber in your lab.

$40,110.61 - $79,749.56

Specifications

Certifications/Compliance UL, CSA, CE
For Use With (Application) Manipulation of anaerobic materials at factory
Voltage 120 V
View More Specs

Products 2
Catalog Number Mfr. No. Depth (English) Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Dimensions (L x W x H) Price Quantity & Availability  
11391106
View Documents Promotion Details
Plas Labs™
855AC/EXP/CN
35 in. 35 x 55 x 38 in. (89 x 140 x 97 cm) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
11386144
View Documents Promotion Details
Plas Labs™
855 ACB
47 in. 119 x 193 x 107 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Illuminated, rigid acrylic glove box with transfer compartment, for manipulation of anaerobic materials at factory-set temperature of 37°C (98.6°F).

General Features

  • Equipped with two pressure/vacuum diaphragm pumps, catalyst heater/oxygen remover, desiccant train and system of purge valves for adjusting interior atmosphere
  • Thick, clear acrylic walls are essentially impermeable to ambient oxygen
  • Useable on a lab cart or table
  • All controls are illuminated

Work Chamber

  • Consists of clear acrylic hood and white thermoset base which clamp together against a double neoprene gasket; clamps adjust for wear
  • Includes two ground key cock valves for purging (not for use with oxygen)
  • Superstructure contains a fluorescent light system and two pumps: one recirculates chamber atmosphere through the desiccant train; the other is for evacuating and purging the transfer compartment and/or work chamber
Transfer Compartment
  • Equipped with two ground key cock purge valves and vacuum gauge
  • Inner door, when unlocked, is self-opening upon equilibration of internal pressure
  • A leveling tray fits bottom of compartment to assist loading and unloading of liquids
Desiccant Cartridges
  • Three clear polycarbonate desiccant cartridges (supplied) are attached in series externally on back of unit
  • They can be disconnected quickly from their self-sealing hose connections without disturbance to the internal atmosphere
  • Each cartridge contains molecular sieve and silica gel medium
  • Contents can be rejuvenated by heating at 163°C (325°F)

Order Info

Electrical Requirements: 120V 60Hz

Certifications

UL listed, CSA certified, CE marked

Specifications

Specifications

UL, CSA, CE
Manipulation of anaerobic materials at factory
120 V
Videos
SDS
Documents

Documents