missing translation for 'onlineSavingsMsg'
Learn More

MOBS, 0.2M buffer soln., pH 8.5, Thermo Scientific™

Catalog No. AAJ62203AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62203AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62203AE Supplier Thermo Scientific Chemicals Supplier No. J62203AE

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 115724-21-5
Color Colorless
Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Molecular Formula C8H17NO4S
MDL Number MFCD00674916
Synonym 4-(4-Morpholinyl)butanesulfonic acid
InChI Key VTOWJTPBPWTSMK-UHFFFAOYSA-N
SMILES OS(=O)(=O)CCCCN1CCOCC1
IUPAC Name 4-(morpholin-4-yl)butane-1-sulfonic acid
Molecular Weight (g/mol) 223.29
Show More Show Less