Learn More
MilliporeSigma™ Supelco™ Mobile Phase Reservoir System
This convenient system is made entirely of inert parts: glass, Tefzel, PEEK, and PTFE TFE. Includes 10 ft./3 m of 1/8 in. O.D. PTFE tubing, a PEEK bottom-of-the-bottle filter/sparger, a plug, and a 1-liter or 2-liter safety-coated bottle.
$510.69 - $769.47
Specifications
For Use With (Application) HPLC
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
111004230
View Documents Promotion Details
MilliporeSigma™ Supelco™
55060U
1 L
Each for $510.69
Only null left
111003931
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA55061
2 L
Each for $769.47
Only null left
Description
Description
Specifications
Specifications
HPLC