missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ MLS 1547
Description
MLS 1547 is a G protein-biased D2 receptor partial agonist (Ki = 1.2 μM). Stimulates D2 receptor G protein-mediated signaling and acts as an antagonist of dopamine-mediated β-arrestin recruitment at D2 receptors.
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-7-[[4-(2-pyridinyl)-1-piperazinyl]methyl]-8-quinolinol |
| CAS | 315698-36-3 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C19H19ClN4O |
| Formula Weight | Observed MW: 354.83 |